2-Bromo-3-methyl-5-(hydroxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

21806-5G 21806-25G 21806-1G 21806-500MG
77632-032EA 749 USD
77632-032 77654-196 77641-248 77649-298
2-Bromo-3-methyl-5-(hydroxymethyl)pyridine ≥97%
2-Bromo-3-methyl-5-(hydroxymethyl)pyridine

Precaución: For Research Use Only

Formula: 202.049 g/mol
MW: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD19105388
CAS Number: 1310416-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR