6-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

21795-5G 21795-1G 21795-500MG 21795-25G
77632-030EA 648 USD
77632-030 77637-238 77652-486 77654-194
6-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester ≥97%
6-Bromo-3-chloro-2-fluorophenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 335.405 g/mol
MW: C₁₂H₁₄BBrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD20526375
CAS Number: 1451391-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR