1-(3,5-Diethylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

216734-5G 216734-1G 216734-25G 216734-500MG
77628-108EA 1513.07 USD
77628-108 77637-234 77654-192 77649-292
1-(3,5-Diethylphenyl)-1-propanone ≥95%
1-(3,5-Diethylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 190.28 g/mol
MW: C₁₃H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD28809603
CAS Number: 1806363-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR