1,3-Diethyl-5-(ethylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

216730-5G 216730-25G 216730-1G 216730-500MG
77628-106EA 1484.23 USD
77628-106 77658-018 77641-242 77649-290
1,3-Diethyl-5-(ethylthio)benzene ≥95%
1,3-Diethyl-5-(ethylthio)benzene

Precaución: For Research Use Only

Formula: 194.34 g/mol
MW: C₁₂H₁₈S
Storage Temperature: Ambient
MDL Number: MFCD22484324
CAS Number: 1824662-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR