1,3-Diethyl-5-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

216684-5G 216684-25G 216684-1G 216684-500MG
77628-104EA 1484.23 USD
77628-104 77654-190 77641-240 77649-288
1,3-Diethyl-5-(trimethylsilyl)benzene ≥95%
1,3-Diethyl-5-(trimethylsilyl)benzene

Precaución: For Research Use Only

Formula: 206.4 g/mol
MW: C₁₃H₂₂Si
Storage Temperature: Ambient
MDL Number: MFCD34868228
CAS Number: 18412-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR