2,3-Dimethyl-6-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

21664-5G 21664-1G 21664-25G 21664-500MG
77632-026EA 656 USD
77632-026 77637-232 77658-016 77645-220
2,3-Dimethyl-6-fluoropyridine ≥97%
2,3-Dimethyl-6-fluoropyridine

Precaución: For Research Use Only

Formula: C₇H₈FN
Storage Temperature: Ambient
MDL Number: MFCD16610908
CAS Number: 1227502-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR