B-(2,4,6-Triethylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

216630-5G 216630-1G 216630-500MG
77654-188EA 1961 USD
77654-188 77641-238 77649-286
B-(2,4,6-Triethylphenyl)boronic acid ≥95%
B-(2,4,6-Triethylphenyl)boronic acid

Precaución: For Research Use Only

Formula: 206.09 g/mol
MW: C₁₂H₁₉BO₂
Storage Temperature: Ambient
MDL Number: MFCD24232458
CAS Number: 108593-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR