2-Fluoro-4-(trifluoromethyl)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid

216349-5G 216349-25G
77634-382EA 148 USD
77634-382 77658-014
2-Fluoro-4-(trifluoromethyl)nicotinic acid ≥97%
2-Fluoro-4-(trifluoromethyl)nicotinic acid

Precaución: For Research Use Only

Formula: C₇H₃F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD10687403
CAS Number: 1040681-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR