3-Bromo-4-chloro-2-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

21631-5G 21631-1G 21631-500MG 21631-25G
77632-024EA 958 USD
77632-024 77637-230 77652-482 77658-012
3-Bromo-4-chloro-2-methoxypyridine ≥95%
3-Bromo-4-chloro-2-methoxypyridine

Precaución: For Research Use Only

Formula: 222.467 g/mol
MW: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD18257595
CAS Number: 1211524-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR