3,4-Difluoro-2-(4-methylphenoxy)-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21542-5G 21542-25G 21542-1G 21542-500MG
77632-022EA 1401 USD
77632-022 77654-186 77641-236 77649-284
3,4-Difluoro-2-(4-methylphenoxy)-benzaldehyde ≥97%
3,4-Difluoro-2-(4-methylphenoxy)-benzaldehyde

Precaución: For Research Use Only

Formula: 248.225 g/mol
MW: C₁₄H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26391664
CAS Number: 2276109-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR