2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

21528-5G 21528-25G 21528-1G 21528-500MG
77632-020EA 808 USD
77632-020 77654-184 77641-234 77645-216
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine ≥95%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-iodopyridine

Precaución: For Research Use Only

Formula: C₁₁H₁₁IN₂
MW: 298.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02186028
CAS Number: 477889-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR