1-Bromo-4-chloro-3-ethynyl-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

214955-1G 214955-5G 214955-500MG
77637-224EA 662 USD
77637-224 77654-178 77645-210
1-Bromo-4-chloro-3-ethynyl-2-fluorobenzene ≥95%
1-Bromo-4-chloro-3-ethynyl-2-fluorobenzene

Precaución: For Research Use Only

Formula: C₈H₃BrClF
Storage Temperature: Ambient
MDL Number: MFCD29037272
CAS Number: 1936019-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR