4-Chloro-1-ethynyl-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

214943-5G 214943-25G
77634-380EA 178 USD
77634-380 77654-176
4-Chloro-1-ethynyl-2-fluorobenzene ≥95%
4-Chloro-1-ethynyl-2-fluorobenzene

Precaución: For Research Use Only

Formula: C₈H₄ClF
Storage Temperature: Ambient
MDL Number: MFCD08703530
CAS Number: 188472-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR