2-(Benzyloxy)-1-bromo-3-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

21492-5G 21492-25G 21492-1G 21492-500MG
77634-378EA 408 USD
77634-378 77658-010 77641-230 77649-282
2-(Benzyloxy)-1-bromo-3-chlorobenzene ≥97%
2-(Benzyloxy)-1-bromo-3-chlorobenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₀BrClO
MW: 297.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977063
CAS Number: 2244107-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR