4-Ethynyl-2-fluoropyridine ≥95%

Supplier: AOBChem USA

Warning

214809-5G 214809-1G 214809-25G 214809-500MG
77632-016EA 727 USD
77632-016 77637-220 77654-174 77649-280
4-Ethynyl-2-fluoropyridine ≥95%
4-Ethynyl-2-fluoropyridine

Precaución: For Research Use Only

Formula: 121.11 g/mol
MW: C₇H₄FN
Storage Temperature: Ambient
MDL Number: MFCD22191415
CAS Number: 1231192-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR