3,5-Dibromo-2-ethynylpyridine ≥95%

Supplier: AOBChem USA

Warning

214786-5G 214786-1G 214786-500MG
77658-006EA 1855 USD
77658-006 77641-228 77649-278
3,5-Dibromo-2-ethynylpyridine ≥95%
3,5-Dibromo-2-ethynylpyridine

Precaución: For Research Use Only

Formula: C₇H₃Br₂N
MW: 260.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26520382
CAS Number: 1567105-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR