6-Chloro-2-fluoro-3-iodobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

213438-5G 213438-25G 213438-1G 213438-500MG
77628-098EA 1484.23 USD
77628-098 77635-530 77637-214 77649-276
6-Chloro-2-fluoro-3-iodobenzonitrile ≥95%
6-Chloro-2-fluoro-3-iodobenzonitrile

Precaución: For Research Use Only

Formula: 281.45 g/mol
MW: C₇H₂ClFIN
Storage Temperature: Ambient
MDL Number: MFCD34867420
CAS Number: 2168872-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR