3-Bromo-5-methyl-2-(1-methylethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21338-5G 21338-25G 21338-1G 21338-500MG
77634-376EA 536 USD
77634-376 77654-170 77641-226 77649-274
3-Bromo-5-methyl-2-(1-methylethoxy)benzaldehyde ≥97%
3-Bromo-5-methyl-2-(1-methylethoxy)benzaldehyde

Precaución: For Research Use Only

Formula: 257.124 g/mol
MW: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD30529797
CAS Number: 1782569-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR