4-Chloro-2-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

21314-5G 21314-1G 21314-500MG
77658-002EA 2522 USD
77658-002 77641-224 77645-204
4-Chloro-2-(trifluoromethoxy)phenylboronic acid ≥95%
4-Chloro-2-(trifluoromethoxy)phenylboronic acid

Precaución: For Research Use Only

Formula: 240.372 g/mol
MW: C₇H₅BClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD12407144
CAS Number: 1095990-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR