Ethyl 6-methoxy-2-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

21301-5G 21301-1G 21301-25G 21301-500MG
77632-012EA 1061 USD
77632-012 77637-212 77658-000 77649-272
Ethyl 6-methoxy-2-methylpyridine-3-carboxylate ≥97%
Ethyl 6-methoxy-2-methylpyridine-3-carboxylate

Precaución: For Research Use Only

Formula: 195.215 g/mol
MW: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD16610988
CAS Number: 173261-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR