6-Ethoxy-2-methylpyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

21298-5G 21298-25G 21298-1G 21298-500MG
77632-010EA 942 USD
77632-010 77657-998 77641-222 77649-270
6-Ethoxy-2-methylpyridine-3-carboxaldehyde ≥97%
6-Ethoxy-2-methylpyridine-3-carboxaldehyde

Precaución: For Research Use Only

Formula: 165.189 g/mol
MW: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD18447617
CAS Number: 1266253-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR