2-Chloro-4-fluoro-3-iodobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

212749-5G 212749-25G 212749-1G 212749-500MG
77632-008EA 1045 USD
77632-008 77657-996 77641-220 77649-268
2-Chloro-4-fluoro-3-iodobenzonitrile ≥95%
2-Chloro-4-fluoro-3-iodobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₂ClFIN
Storage Temperature: Ambient
MDL Number: MFCD30180182
CAS Number: 899821-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR