2,3-Dichloro-4-fluorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

212748-5G 212748-25G 212748-1G
77634-374EA 223 USD
77634-374 77657-994 77641-218
2,3-Dichloro-4-fluorobenzonitrile ≥95%
2,3-Dichloro-4-fluorobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₂Cl₂FN
MW: 190.0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690560
CAS Number: 908123-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR