2-Fluoro-5-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21231-5G 21231-25G 21231-1G 21231-500MG
77634-372EA 548 USD
77634-372 77657-990 77641-214 77649-266
2-Fluoro-5-isopropoxybenzaldehyde ≥97%
2-Fluoro-5-isopropoxybenzaldehyde

Precaución: For Research Use Only

Formula: 182.192 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD24685709
CAS Number: 219939-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR