3-Bromo-4-fluoro-1-cyclopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

212153-5G 212153-25G 212153-1G 212153-500MG
77634-370EA 361 USD
77634-370 77657-986 77641-212 77649-260
3-Bromo-4-fluoro-1-cyclopropylbenzene ≥95%
3-Bromo-4-fluoro-1-cyclopropylbenzene

Precaución: For Research Use Only

Formula: C₉H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD21099995
CAS Number: 1353854-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR