2-Methyl-5-phenyl-1,3,4-oxadiazole ≥97%

Supplier: AOBChem USA

Warning

21214-5G 21214-1G 21214-25G 21214-500MG
77632-004EA 935 USD
77632-004 77637-206 77654-166 77649-258
2-Methyl-5-phenyl-1,3,4-oxadiazole ≥97%
2-Methyl-5-phenyl-1,3,4-oxadiazole

Precaución: For Research Use Only

Formula: 160.173 g/mol
MW: C₉H₈N₂O
Storage Temperature: Ambient
CAS Number: 4046-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR