2-Bromo-4-fluoro-1-cyclopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

212076-5G 212076-1G 212076-500MG 212076-25G
77634-368EA 479 USD
77634-368 77637-204 77652-478 77657-984
2-Bromo-4-fluoro-1-cyclopropylbenzene ≥97%
2-Bromo-4-fluoro-1-cyclopropylbenzene

Precaución: For Research Use Only

Formula: C₉H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD21100009
CAS Number: 1353855-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR