3,4-Difluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21169-5G 21169-1G 21169-25G 21169-500MG
77634-366EA 579 USD
77634-366 77637-202 77654-164 77645-202
3,4-Difluoro-2-methoxybenzaldehyde ≥97%
3,4-Difluoro-2-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115951
CAS Number: 1023023-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR