2-Bromo-3-fluorobenzal fluoride ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-(difluoromethyl)-3-fluorobenzene

211014-5G 211014-25G 211014-1G 211014-500MG
77634-362EA 571 USD
77634-362 77654-160 77641-208 77649-256
2-Bromo-3-fluorobenzal fluoride ≥95%
2-Bromo-3-fluorobenzal fluoride

Precaución: For Research Use Only

Formula: C₇H₄BrF₃
MW: 225.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698531
CAS Number: 1214338-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR