2-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210435-5G 210435-1G 210435-25G 210435-500MG
77628-094EA 1484.23 USD
77628-094 77637-198 77654-158 77649-254
2-Ethoxy-α-(1-ethylpropyl)benzenemethanol ≥95%
2-Ethoxy-α-(1-ethylpropyl)benzenemethanol

Precaución: For Research Use Only

Formula: 222.32 g/mol
MW: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19601795
CAS Number: 1339742-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR