α-(1-Ethylpropyl)-4-fluoro-3-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210327-5G 210327-25G 210327-1G 210327-500MG
77628-092EA 1484.23 USD
77628-092 77635-528 77641-202 77649-250
α-(1-Ethylpropyl)-4-fluoro-3-methoxybenzenemethanol ≥95%
α-(1-Ethylpropyl)-4-fluoro-3-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₁₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34867089
CAS Number: 1596953-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR