1,3-Dibromo-5-ethyl-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

209855-5G 209855-1G 209855-25G 209855-500MG
77628-090EA 1484.23 USD
77628-090 77637-194 77654-154 77649-246
1,3-Dibromo-5-ethyl-2-fluorobenzene ≥95%
1,3-Dibromo-5-ethyl-2-fluorobenzene

Precaución: For Research Use Only

Formula: C₈H₇Br₂F
Storage Temperature: Ambient
MDL Number: MFCD28784544
CAS Number: 1807055-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR