2-Bromo-1,3,4,5-tetrafluorobenzene ≥95%

Supplier: AOBChem USA

Warning

209818-5G 209818-25G 209818-1G 209818-500MG
77628-088EA 1484.23 USD
77628-088 77657-974 77641-200 77645-196
2-Bromo-1,3,4,5-tetrafluorobenzene ≥95%
2-Bromo-1,3,4,5-tetrafluorobenzene

Precaución: For Research Use Only

Formula: C₆HBrF₄
MW: 228.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000303
CAS Number: 1559-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR