1-Bromo-2,4,5-trifluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

209760-5G 209760-1G 209760-500MG 209760-25G
77631-998EA 1084 USD
77631-998 77637-192 77652-470 77657-972
1-Bromo-2,4,5-trifluoro-3-methoxybenzene ≥97%
1-Bromo-2,4,5-trifluoro-3-methoxybenzene

Precaución: For Research Use Only

Formula: C₇H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD25958094
CAS Number: 13332-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR