4-Bromotetrafluorophenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2,3,5,6-tetrafluorophenol

209713-5G 209713-25G
77634-354EA 59 USD
77634-354 77657-968
4-Bromotetrafluorophenol ≥97%
4-Bromotetrafluorophenol

Precaución: For Research Use Only

Formula: C₆HBrF₄O
MW: 244.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246024
CAS Number: 1998-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR