4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

209710-5G 209710-25G
77634-352EA 74 USD
77634-352 77654-150
4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥97%
4-Bromo-2,3,5,6-tetrafluorobenzoic acid

Precaución: For Research Use Only

Formula: C₇HBrF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00075244
CAS Number: 4707-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR