Methyl 4-bromo-2,3,5,6-tetrafluorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
209683-5G
209683-25G
77634-350EA
123
USD
77634-350
77657-966
Methyl 4-bromo-2,3,5,6-tetrafluorobenzoate ≥97%
Methyl 4-bromo-2,3,5,6-tetrafluorobenzoate
Precaución: For Research Use Only
Formula:
C₈H₃BrF₄O₂ MW: 287.0058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14698132 CAS Number: 4707-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...