4-Iodo-6-methoxy-3-pyridinol ≥95%

Supplier: AOBChem USA

Warning

209537-5G 209537-1G 209537-500MG 209537-25G
77631-996EA 1368 USD
77631-996 77637-190 77652-468 77657-964
4-Iodo-6-methoxy-3-pyridinol ≥95%
4-Iodo-6-methoxy-3-pyridinol

Precaución: For Research Use Only

Formula: 251.02 g/mol
MW: C₆H₆INO₂
Storage Temperature: Ambient
MDL Number: MFCD14705086
CAS Number: 431942-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR