4-[4-(Trifluoromethoxy)phenoxy]benzonitrile ≥97%

Supplier: AOBChem USA

Warning

209191-5G 209191-25G 209191-1G 209191-500MG
77628-086EA 1484.23 USD
77628-086 77635-526 77637-188 77645-194
4-[4-(Trifluoromethoxy)phenoxy]benzonitrile ≥97%
4-[4-(Trifluoromethoxy)phenoxy]benzonitrile

Precaución: For Research Use Only

Formula: C₁₄H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD26517596
CAS Number: 1258834-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR