1-(2,3,5,6-Tetrafluorophenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

208963-5G 208963-500MG 208963-25G 208963-1G
77631-994EA 1357 USD
77631-994 77652-466 77654-148 77641-196
1-(2,3,5,6-Tetrafluorophenyl)-1-propanone ≥95%
1-(2,3,5,6-Tetrafluorophenyl)-1-propanone

Precaución: For Research Use Only

Formula: 206.14 g/mol
MW: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD34866805
CAS Number: 135762-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR