1-Bromo-2-chloro-4-methylsulfanyl-benzene ≥95%

Supplier: AOBChem USA

Warning

20895-1G 20895-5G 20895-500MG
77637-186EA 685 USD
77637-186 77654-146 77649-242
1-Bromo-2-chloro-4-methylsulfanyl-benzene ≥95%
1-Bromo-2-chloro-4-methylsulfanyl-benzene

Precaución: For Research Use Only

Formula: 237.545 g/mol
MW: C₇H₆BrClS
Storage Temperature: Ambient
MDL Number: MFCD22488865
CAS Number: 101083-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR