2-Bromo-5-fluoronicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

20883-5G 20883-25G 20883-1G 20883-500MG
77634-348EA 428 USD
77634-348 77657-962 77641-194 77649-240
2-Bromo-5-fluoronicotinaldehyde ≥97%
2-Bromo-5-fluoronicotinaldehyde

Precaución: For Research Use Only

Formula: C₆H₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD16606507
CAS Number: 1227562-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR