2-Iodo-4,5-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

20778-5G 20778-25G 20778-1G 20778-500MG
77631-990EA 835 USD
77631-990 77657-956 77641-190 77649-232
2-Iodo-4,5-dimethoxybenzaldehyde ≥95%
2-Iodo-4,5-dimethoxybenzaldehyde

Precaución: For Research Use Only

Formula: 292.07 g/mol
MW: C₉H₉IO₃
Storage Temperature: Ambient
MDL Number: MFCD13193552
CAS Number: 61203-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR