2-Chloro-5-methoxymethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

20746-5G 20746-1G 20746-25G 20746-500MG
77631-988EA 648 USD
77631-988 77637-180 77654-144 77645-192
2-Chloro-5-methoxymethylphenylboronic acid pinacol ester ≥97%
2-Chloro-5-methoxymethylphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₂₀BClO₃
Storage Temperature: Ambient
MDL Number: MFCD18730001
CAS Number: 1079402-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR