1,2-Benzenediol, 3-bromo-6-chloro- ≥97%

Supplier: AOBChem USA

Warning

207429-5G 207429-1G 207429-25G 207429-500MG
77628-080EA 1484.23 USD
77628-080 77637-178 77657-954 77645-190
1,2-Benzenediol, 3-bromo-6-chloro- ≥97%
1,2-Benzenediol, 3-bromo-6-chloro-

Precaución: For Research Use Only

Formula: 223.45 g/mol
MW: C₆H₄BrClO₂
Storage Temperature: Ambient
CAS Number: 180268-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR