1,3-Dibromo-2-butoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

20726-5G 20726-1G 20726-25G 20726-500MG
77634-346EA 378 USD
77634-346 77637-176 77657-952 77645-188
1,3-Dibromo-2-butoxy-5-methylbenzene ≥97%
1,3-Dibromo-2-butoxy-5-methylbenzene

Precaución: For Research Use Only

Formula: 322.036 g/mol
MW: C₁₁H₁₄Br₂O
Storage Temperature: Ambient
MDL Number: MFCD17015813
CAS Number: 1245563-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR