5-Bromo-4-fluoropyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

20721-5G 20721-25G
77634-344EA 59 USD
77634-344 77654-142
5-Bromo-4-fluoropyridin-2-amine ≥97%
5-Bromo-4-fluoropyridin-2-amine

Precaución: For Research Use Only

Formula: C₅H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD16658678
CAS Number: 944401-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR