2-Ethyl-3-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

207066-1G 207066-5G 207066-500MG
77634-342EA 1198 USD
77634-342 77657-950 77645-186
2-Ethyl-3-fluorobenzonitrile ≥97%
2-Ethyl-3-fluorobenzonitrile

Precaución: For Research Use Only

Formula: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD22490763
CAS Number: 1369938-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR