5-Bromo-2-methyl-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

206324-5G 206324-1G 206324-25G 206324-500MG
77631-984EA 927 USD
77631-984 77637-172 77657-946 77645-180
5-Bromo-2-methyl-3-(trifluoromethyl)aniline ≥97%
5-Bromo-2-methyl-3-(trifluoromethyl)aniline

Precaución: For Research Use Only

Formula: C₈H₇BrF₃N
MW: 254.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28746066
CAS Number: 1804407-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR