5-Bromo-2-methyl-1-nitro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

206310-500MG 206310-5G 206310-1G
77652-464EA 386 USD
77652-464 77657-944 77641-188
5-Bromo-2-methyl-1-nitro-3-(trifluoromethyl)benzene ≥97%
5-Bromo-2-methyl-1-nitro-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₈H₅BrF₃NO₂
Storage Temperature: Ambient
CAS Number: 1698027-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR